Learn More
Biomatik Corporation Mouse Platelet Factor 4,PF-4 ELISA Kit
Supplier:  Biomatik Corporation EKC31418
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.125 ng/mL-200 ng/mL. Sensitivity: 1.603 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9Z126. Target Synonyms: CXCL4, MGC138298, SCYB4, chemokine (C-X-C motif) ligand 4|platelet factor 4 (chemokine (C-X-C motif) ligand 4). Restriction: For Research Use Only.
Catalog No. 50-229-3607