Learn More
Biomatik Corporation Pig Albumin (Alb) ELISA Kit
Supplier:  Biomatik Corporation EKC31440
Encompass_Preferred
Sample Type: serum, plasma, urine. Species Reactivity: Sus scrofa (Pig). Detection Range: 0.03 μg/mL-7000 μg/mL. Sensitivity: 0.03 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P08835. Target Synonyms: DKFZp779N1935, PRO0883, PRO0903, PRO1341, cell growth inhibiting protein 42|growth-inhibiting protein 20|serum albumin. Restriction: For Research Use Only.
Catalog No. 50-229-3611