Learn More
Biomatik Corporation Pig Free Thyroxine (Ft4) Elisa Kit
Supplier:  Biomatik Corporation EKC31447
Encompass_Preferred
Pig Free Thyroxine (Ft4) Elisa Kit for detecting Free Thyroxine (Ft4) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 4-64pmol/l. Sensitivity: 1.08pmol/l. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3613