Learn More
Biomatik Corporation Rabbit apolipoprotein B100 (Apo-B100) ELISA Kit
Supplier:  Biomatik Corporation EKC31463
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 78 ng/mL-5000 ng/mL. Sensitivity: 19.5 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P17165. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3615