Learn More
Biomatik Corporation Rabbit C-Peptide ELISA kit
Supplier:  Biomatik Corporation EKC31467
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 0.5 ng/mL-15 ng/mL. Sensitivity: 0.3 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01311. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3617