Learn More
Biomatik Corporation Rat Vitamin A,VA ELISA Kit
Supplier:  Biomatik Corporation EKC31502
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 312.5 pg/mL-5000 pg/mL. Sensitivity: 233.598 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3624