Learn More
Biomatik Corporation Bovine Caspase 7 (Casp-7) Elisa Kit
Supplier:  Biomatik Corporation EKC31578
Encompass_Preferred
Bovine Caspase 7 (Casp-7) Elisa Kit for detecting Caspase 7(Casp-7) in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: F1MD58; CASP7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3632