Learn More
Biomatik Corporation Bovine Lipopolysaccharide Binding Protein (Lbp) Elisa Kit
Supplier:  Biomatik Corporation EKC31716
Encompass_Preferred
Bovine Lipopolysaccharide Binding Protein (Lbp) Elisa Kit for detecting Lipopolysaccharide Binding Protein in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q2TBI0; LBP. Target Synonyms: LBPLipopolysaccharide-binding protein; LBP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3645