Learn More
Biomatik Corporation Bovine Selenoprotein P (Sepp1) Elisa Kit
Supplier:  Biomatik Corporation EKC31785
Encompass_Preferred
Bovine Selenoprotein P (Sepp1) Elisa Kit for detecting Selenoprotein P, Plasma, 1 in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 10-160ng/ml. Sensitivity: 5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P49907; SELENOP. Target Synonyms: SELENOP; Selenoprotein P; SeP; Selenoprotein P-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3654