Learn More
Biomatik Corporation Canine Calprotectin (Calp) Elisa Kit
Supplier:  Biomatik Corporation EKC31838
Encompass_Preferred
Canine Calprotectin (Calp) Elisa Kit for detecting Calprotectin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3662