Learn More
Biomatik Corporation Canine Follicle-Stimulating Hormone, Fsh Elisa Kit
Supplier:  Biomatik Corporation EKC31850
Encompass_Preferred
Canine Follicle-Stimulating Hormone, Fsh Elisa Kit for detecting Follicle-Stimulating Hormone, Fsh in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.35-140mIU/ml. Sensitivity: 0.35mIU/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: E2RJZ8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3666