Learn More
Biomatik Corporation Canine Kidney Injury Molecule 1 (Kim-1) Elisa Kit
Supplier:  Biomatik Corporation EKC31861
Encompass_Preferred
Canine Kidney Injury Molecule 1 (Kim-1) Elisa Kit for detecting Kidney Injury Molecule 1(Kim-1) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3668