Learn More
Biomatik Corporation Canine Matrix Metalloproteinase 2/Gelatinase A, Mmp-2 Elisa Kit
Supplier:  Biomatik Corporation EKC31867
Encompass_Preferred
Canine Matrix Metalloproteinase 2/Gelatinase A, Mmp-2 Elisa Kit for detecting Matrix Metalloproteinase 2/Gelatinase A, Mmp-2 in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 4.69-300ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: F1PMK7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3671