Learn More
Biomatik Corporation Chicken Heat Shock Protein 90, Hsp-90 Elisa Kit
Supplier:  Biomatik Corporation EKC31937
Encompass_Preferred
Chicken Heat Shock Protein 90, Hsp-90 Elisa Kit for detecting Heat Shock Protein 90Kda Alpha (Cytosolic), Class A Member 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Chicken (Gallus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11501; HSP90AA1. Target Synonyms: HSP90AA1; HSPCAHeat shock protein HSP 90-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3683