Learn More
Biomatik Corporation Chicken Peptide Yy (Pyy) Elisa Kit
Supplier:  Biomatik Corporation EKC31977
Encompass_Preferred
Chicken Peptide Yy (Pyy) Elisa Kit for detecting Peptide Yy in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 50-800pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P29203; PYY. Target Synonyms: PYY; Peptide YY-like; PYY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3689