Learn More
Biomatik Corporation Dog angiotensin Ⅱ (ANG-Ⅱ) ELISA Kit
Supplier:  Biomatik Corporation EKC32006
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Canis lupus familiaris (Dog) (Canis familiaris). Detection Range: 6.25 pg/mL-400 pg/mL. Sensitivity: 1.56 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: angiotension Ⅱ, angiotensin 2, ANG2. Restriction: For Research Use Only.
Catalog No. 50-229-3692