Learn More
Biomatik Corporation Dog Circulating Immune Complex (Cic) Elisa Kit
Supplier:  Biomatik Corporation EKC32026
Encompass_Preferred
Dog Circulating Immune Complex (Cic) Elisa Kit for detecting Circulating Immune Complex (Cic) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 2.34-1200ng/ml. Sensitivity: 2.34ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3694