Learn More
Biomatik Corporation Dog Insulin-Like Growth Factor 1 (Igf1) Elisa Kit
Supplier:  Biomatik Corporation EKC32059
Encompass_Preferred
Dog Insulin-Like Growth Factor 1 (Igf1) Elisa Kit for detecting Insulin-Like Growth Factor 1 (Somatomedin C) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 31.25-2000ng/ml. Sensitivity: 7.8ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P33712; IGF1. Target Synonyms: IGF1; IGFIAInsulin-like growth factor I; IGF-I; Somatomedin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3698