Learn More
Biomatik Corporation Dog Interleukin-33 (Il33) Elisa Kit
Supplier:  Biomatik Corporation EKC32082
Encompass_Preferred
Dog Interleukin-33 (Il33) Elisa Kit for detecting Interleukin 33 in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 23.44-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O97863; IL33. Target Synonyms: IL33Interleukin-33; IL-33; Protein DVS27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3702