Learn More
Biomatik Corporation Fish Aromatase (Aro) Elisa Kit
Supplier:  Biomatik Corporation EKC32144
Encompass_Preferred
Fish Aromatase (Aro) Elisa Kit for detecting Cytochrome P450, Family 19, Subfamily A, Polypeptide 1 in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 25-1000pg/ml. Sensitivity: 25pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q92087; cyp19a1. Target Synonyms: cyp19a1; cyp19Aromatase; EC 1.14.14.14; CYPXIX; Cytochrome P-450AROM; Cytochrome P450 19A1; Estrogen synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3713