Learn More
Biomatik Corporation Goat follicle-stimulating hormone,FSH ELISA Kit
Supplier:  Biomatik Corporation EKC32191
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Capra hircus (Goat). Detection Range: 20 mIU/mL-800 mIU/mL. Sensitivity: 20 mIU/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8HY83. Target Synonyms: follicle-stimulating hormone beta subunit|follitropin, beta chain. Restriction: For Research Use Only.
Catalog No. 50-229-3719