Learn More
Biomatik Corporation Goat Growth Hormone, Gh Elisa Kit
Supplier:  Biomatik Corporation EKC32197
Encompass_Preferred
Goat Growth Hormone, Gh Elisa Kit for detecting Growth Hormone 1 in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 6.25-100ng/ml. Sensitivity: 6.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P67931; GH1. Target Synonyms: GH1; Somatotropin; Growth hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3721