Learn More
Biomatik Corporation Goat Insulin-Like Growth Factor 1 (Igf-1) Elisa Kit
Supplier:  Biomatik Corporation EKC32204
Encompass_Preferred
Goat Insulin-Like Growth Factor 1 (Igf-1) Elisa Kit for detecting Insulin-Like Growth Factor 1 (Somatomedin C) in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 80-1000ng/ml. Sensitivity: 20ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51457; IGF1. Target Synonyms: IGF1Insulin-like growth factor I; IGF-I; Somatomedin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3723