Learn More
Biomatik Corporation Goat Thyroid Stimulating Hormone, Tsh Elisa Kit
Supplier:  Biomatik Corporation EKC32232
Encompass_Preferred
Goat Thyroid Stimulating Hormone, Tsh Elisa Kit for detecting Glycoprotein Hormones, Alpha Polypeptide in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 0.5-20uIU/ml. Sensitivity: 0.25uIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8WMW8; CGA. Target Synonyms: CGAGlycoprotein hormones alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha; Follicle-stimulating hormone alpha chain; FSH-alpha; Follitropin alpha chain; Luteinizing hormone alpha chain; LSH-alpha; Lutropin alpha chain; Thyroid-stimulating hormone alpha chain; TSH-alpha; Thyrotropin alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3727