Learn More
Biomatik Corporation Hamster Interleukin-4(IL4) ELISA kit
Supplier:  Biomatik Corporation EKC32295
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mesocricetus auratus (Golden hamster). Detection Range: 9.37 pg/mL-600 pg/mL. Sensitivity: 2.34 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q60440. Target Synonyms: BCGF-1, BCGF1, BSF1, IL-4, MGC79402, B cell growth factor 1|B_cell stimulatory factor 1|lymphocyte stimulatory factor 1. Restriction: For Research Use Only.
Catalog No. 50-229-3733