Learn More
Biomatik Corporation Horse Gelsolin (Gsn) Elisa Kit
Supplier:  Biomatik Corporation EKC32326
Encompass_Preferred
Horse Gelsolin (Gsn) Elisa Kit for detecting Gelsolin (Amyloidosis, Finnish Type) in serum, plasma, tissue homogenates. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 12.5-800ng/ml. Sensitivity: 6.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q28372; GSN. Target Synonyms: GSNGelsolin; Actin-depolymerizing factor; ADF; Brevin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3738