Learn More
Biomatik Corporation Horse Hepcidin (Hamp) Elisa Kit
Supplier:  Biomatik Corporation EKC32332
Encompass_Preferred
Horse Hepcidin (Hamp) Elisa Kit for detecting Hepcidin Antimicrobial Peptide in serum, plasma, tissue homogenates. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 7.8-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: HEPC; HEPCIDIN; HFE2B; LEAP-1; LEAP1; PLTR; liver-expressed antimicrobial peptide; putative liver tumor regressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3740