Learn More
Biomatik Corporation Horse Insulin-Like Growth Factor 1 (Igf1) Elisa Kit
Supplier:  Biomatik Corporation EKC32337
Encompass_Preferred
Horse Insulin-Like Growth Factor 1 (Igf1) Elisa Kit for detecting Insulin-Like Growth Factor 1 (Somatomedin C) in serum, plasma, tissue homogenates. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 12.5-800ng/ml. Sensitivity: 3.12ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51458; IGF1. Target Synonyms: IGF1; Insulin-like growth factor I; IGF-I; Somatomedin; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3743