Learn More
Biomatik Corporation Human 14-3-3 Protein Eta (Ywhah) Elisa Kit
Supplier:  Biomatik Corporation EKC32382
Encompass_Preferred
Human 14-3-3 Protein Eta (Ywhah) Elisa Kit for detecting Tyrosine 3-Monooxygenase/Tryptophan 5-Monooxygenase Activation Protein, Eta Polypeptide in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q04917; YWHAH. Target Synonyms: 14 3 3 protein eta; 14-3-3 eta; 14-3-3 protein eta; 1433F_HUMAN; Brain protein 14-3-3; eta isoform; HGNC:12853; Protein AS1; Tyrosine 3 monooxygenase/tryptophan 5 monooxygenase activation protein 1; Tyrosine 3 monooxygenase/tryptophan 5 monooxygenase activation protein eta polypeptide; Tyrosine 3 monooxygenase/tryptophan 5 monooxygenase activation protein; eta isoform; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein; eta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3749