Learn More
Biomatik Corporation Human 3-hydroxyacyl-CoA dehydrogenase type-2(HSD17B10) ELISA kit
Supplier:  Biomatik Corporation EKC32400
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 15.625 pg/ml- 1000pg/ml. Sensitivity: 3.9 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q99714. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3755