Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 33, Adam33 Elisa Kit
Supplier:  Biomatik Corporation EKC32429
Encompass_Preferred
Human A Disintegrin And Metalloprotease 33, Adam33 Elisa Kit for detecting Adam Metallopeptidase Domain 33 in serum, cell culture supernates, saliva, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9BZ11; ADAM33. Target Synonyms: A disintegrin and metalloprotease 33; A disintegrin and metalloproteinase domain 33; ADA33_HUMAN; ADAM 33; ADAM metallopeptidase domain 33; ADAM33; C20orf153; Disintegrin and metalloproteinase domain-containing protein 33; Disintegrin and reprolysin metalloproteinase family protein; DJ964F7.1; DKFZp434K0521; FLJ35308; FLJ36751; Metalloprotease disintegrin; MGC149823; MGC71889; PRO1891; UNQ873. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3761