Learn More
Biomatik Corporation Human Cystatin-F (CST7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33353
Encompass_Preferred
The Human Cystatin-F (CST7) ELISA Kit allows for quantitative determination of cystatin F (leukocystatin) in serum, plasma, cell culture supernates. Species Reactivity: Human. Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O76096. Restriction: For Research Use Only.
Catalog No. 50-149-7824