Learn More
Biomatik Corporation Human a proliferation inducing ligand(APRIL)ELISA Kit
Supplier:  Biomatik Corporation EKC32450
Encompass_Preferred
Sample Type: serum, plasma, urine. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78 ng/mL-50 ng/mL. Sensitivity: 0.195 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O75888. Target Synonyms: APRIL, CD256, TALL2, TRDL-1, UNQ383/PRO715, ligand, TNF- and APOL-related leukocyte expressed ligand 2|a proliferation inducing ligand|tumor necrosis factor (ligand) superfamily member 13 transcript. Restriction: For Research Use Only.
Catalog No. 50-229-3765