Learn More
Biomatik Corporation Human Activated Coagulation Factor Xii, Fxiia Elisa Kit
Supplier:  Biomatik Corporation EKC32471
Encompass_Preferred
Human Activated Coagulation Factor Xii, Fxiia Elisa Kit for detecting Activated Coagulation Factor XII, FXIIa in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3767