Learn More
Biomatik Corporation Human Acylamino-Acid-Releasing Enzyme (Apeh) Elisa Kit
Supplier:  Biomatik Corporation EKC32478
Encompass_Preferred
Human Acylamino-Acid-Releasing Enzyme (Apeh) Elisa Kit for detecting N-Acylaminoacyl-Peptide Hydrolase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13798; APEH. Target Synonyms: AARE; ACPH; ACPH_HUMAN; Acyl peptide hydrolase; Acyl-peptide hydrolase; Acylamino acid releasing enzyme; Acylamino-acid-releasing enzyme; Acylaminoacyl peptidase; Acylaminoacyl-peptidase; APEH; APH; N acylaminoacyl peptide hydrolase; OPH; Oxidized protein hydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3769