Learn More
Biomatik Corporation Human Adam Dec1 (Adamdec1) Elisa Kit
Supplier:  Biomatik Corporation EKC32485
Encompass_Preferred
Human Adam Dec1 (Adamdec1) Elisa Kit for detecting Adam-Like, Decysin 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O15204; ADAMDEC1. Target Synonyms: A disintegrin and metalloproteinase domain like protein decysin 1; A disintegrin and metalloproteinase domain-like protein decysin-1; ADAM DEC1; ADAM like decysin 1; ADAM like protein decysin 1; ADAM-like protein decysin-1; ADAMDEC 1; Adamdec1; ADEC1_HUMAN; Decysin; Disintegrin protease; M12.219. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3771