Learn More
Biomatik Corporation Human Alanyl (Membrane) Aminopeptidase (Anpep) Elisa Kit
Supplier:  Biomatik Corporation EKC32517
Encompass_Preferred
Human Alanyl (Membrane) Aminopeptidase (Anpep) Elisa Kit for detecting Alanyl (Membrane) Aminopeptidase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P15144; ANPEP. Target Synonyms: Alanyl (membrane) aminopeptidase; Alanyl aminopeptidase; Aminopeptidase M; Aminopeptidase N; AMPN_HUMAN; ANPEP; AP M; AP N; AP-M; AP-N; APN; CD 13; CD13; CD13 antigen; gp150; hAPN; LAP 1; LAP1; Microsomal aminopeptidase; Myeloid plasma membrane glycoprotein CD13; p150; PEPN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3782