Learn More
Biomatik Corporation Human Angiotensin Ii Receptor 1 Antibody, Atiir1 Ab Elisa Kit
Supplier:  Biomatik Corporation EKC32589
Encompass_Preferred
Human Angiotensin Ii Receptor 1 Antibody, Atiir1 Ab Elisa Kit for detecting Angiotensin II Receptor 1 Antibody, AtIIr1 Ab in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: angiotensin 2 receptor 1 antibody; AT2R1-Ab. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3793