Learn More
Biomatik Corporation Human Ankyrin Repeat Domain-Containing Protein 26 (Ankrd26) Elisa Kit
Supplier:  Biomatik Corporation EKC32598
Encompass_Preferred
Human Ankyrin Repeat Domain-Containing Protein 26 (Ankrd26) Elisa Kit for detecting Ankyrin Repeat Domain 26 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UPS8; ANKRD26. Target Synonyms: Ankrd26; Ankyrin repeat domain 26; Ankyrin repeat domain-containing protein 26; Ankyrin repeat domaincontaining protein 26; ANR26_HUMAN; bA145E8.1; KIAA1074. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3795