Learn More
Biomatik Corporation Human anti-growth hormone(GH) antibody ELISA Kit
Supplier:  Biomatik Corporation EKC32675
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 2 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3800