Learn More
Biomatik Corporation Human Apolipoprotein O (Apoo) Elisa Kit
Supplier:  Biomatik Corporation EKC32770
Encompass_Preferred
Human Apolipoprotein O (Apoo) Elisa Kit for detecting Apolipoprotein O in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.08ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q9BUR5; APOO. Target Synonyms: APOO; FAM121B; MIC23; MIC26; My025; UNQ1866/PRO4302; MICOS complex subunit MIC26; Apolipoprotein O; MICOS complex subunit MIC23; Protein FAM121B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3807