Learn More
Biomatik Corporation Human Atp Synthase Subunit Beta, Mitochondrial (Atp5B) Elisa Kit
Supplier:  Biomatik Corporation EKC32806
Encompass_Preferred
Human Atp Synthase Subunit Beta, Mitochondrial (Atp5B) Elisa Kit for detecting Atp Synthase, H+ Transporting, Mitochondrial F1 Complex, Beta Polypeptide in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06576; ATP5B. Target Synonyms: ATP 5B; ATP synthase H+ transporting mitochondrial F1 complex beta polypeptide; ATP synthase subunit beta mitochondrial; ATP synthase subunit beta; mitochondrial; atp5b; ATPB; ATPB_HUMAN; ATPMB; ATPSB; Epididymis secretory protein Li 271; HEL-S-271; Mitochondrial ATP synthase beta subunit; Mitochondrial ATP Synthase Subunit Beta; Mitochondrial ATP synthetase beta subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3813