Learn More
Biomatik Corporation Human C4B-Binding Protein Beta Chain (C4Bpb) Elisa Kit
Supplier:  Biomatik Corporation EKC32933
Encompass_Preferred
Human C4B-Binding Protein Beta Chain (C4Bpb) Elisa Kit for detecting Complement Component 4 Binding Protein, Beta in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20851; C4BPB. Target Synonyms: C4b binding protein beta chain; C4b-binding protein beta chain; C4BP; C4BPB; C4BPB_HUMAN; Complement component 4 binding protein beta; Complement component 4 binding protein beta chain; Complement component 4-binding protein beta; OTTHUMP00000034288; OTTHUMP00000034289; OTTHUMP00000034375; OTTHUMP0000003474; Proline rich protein; PRP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3833