Learn More
Biomatik Corporation Human Cartilage Intermediate Layer Protein 2 (Cilp2) Elisa Kit
Supplier:  Biomatik Corporation EKC33021
Encompass_Preferred
Human Cartilage Intermediate Layer Protein 2 (Cilp2) Elisa Kit for detecting Cartilage Intermediate Layer Protein 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8IUL8; CILP2. Target Synonyms: CILP2; Cartilage intermediate layer protein 2; CILP-2) [Cleaved into: Cartilage intermediate layer protein 2 C1; Cartilage intermediate layer protein 2 C2]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3849