Learn More
Biomatik Corporation Human Cathepsin G (Cath-G) Elisa Kit
Supplier:  Biomatik Corporation EKC33044
Encompass_Preferred
Human Cathepsin G (Cath-G) Elisa Kit for detecting Cathepsin G in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08311; CTSG. Target Synonyms: CATG; CATG_HUMAN; Cathepsin G; Cathepsin G precursor; CathepsinG; CG; Ctsg; CTSG protein; MGC23078. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3853