Learn More
Biomatik Corporation Human Cd226 Antigen (Cd226) Elisa Kit
Supplier:  Biomatik Corporation EKC33072
Encompass_Preferred
Human Cd226 Antigen (Cd226) Elisa Kit for detecting Cd226 Molecule in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60ng/ml. Sensitivity: 0.22ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15762; CD226. Target Synonyms: adhesion glycoprotein; CD226; CD226 antigen; CD226 molecule; CD226_HUMAN; DNAM 1; DNAM-1; DNAM1; DNAX accessory molecule 1; platelet and T cell activation antigen 1; PTA1; T lineage specific activation antigen 1 antigen; TLiSA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3858