Learn More
Biomatik Corporation Human Chitinase 1 (Chitotriosidase) (Chit1) Elisa Kit
Supplier:  Biomatik Corporation EKC33110
Encompass_Preferred
Human Chitinase 1 (Chitotriosidase) (Chit1) Elisa Kit for detecting Chitinase 1 (Chitotriosidase) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13231; CHIT1. Target Synonyms: CHI3; CHIT; CHIT1; CHIT1_HUMAN; CHITD; Chitinase 1; Chitinase 1 (chitotriosidase); Chitinase-1; Chitotriosidase; Chitotriosidase-1; EC 3.2.1.14; FLJ00314; MGC125322; Plasma methylumbelliferyl tetra N acetylchitotetraoside hydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3863