Learn More
Biomatik Corporation Human Claudin-7 (Cldn7) Elisa Kit
Supplier:  Biomatik Corporation EKC33148
Encompass_Preferred
Human Claudin-7 (Cldn7) Elisa Kit for detecting Claudin 7 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95471; CLDN7. Target Synonyms: CLDN7; CEPTRL2; CPETRL2; Claudin-7; CLDN-7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3867