Learn More
Biomatik Corporation Human Cluster Of Differentiation, Cd44 Elisa Kit
Supplier:  Biomatik Corporation EKC33169
Encompass_Preferred
Human Cluster Of Differentiation, Cd44 Elisa Kit for detecting Cd44 Molecule (Indian Blood Group) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16070; CD44. Target Synonyms: LHR; BA-1; CD 44; CD44; CD44 antigen; CD44 molecule (Indian blood group); CD44 molecule; CD44_HUMAN; CDw44; CDW44 antigen; Cell surface glycoprotein CD44; chondroitin sulfate proteoglycan 8; CSPG8; ECMR-III; Epican; Extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; HCELL; hematopoietic cell E- and L-selectin ligand; Heparan sulfate proteoglycan; Hermes antigen; homing function and Indian blood group system; HSA; HUTCH-I; HUTCH1; HUTCHI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3872