Learn More
Biomatik Corporation Human Cocaine-And Amphetamine-Regulated Transcript Protein (Cartpt) Elisa Kit
Supplier:  Biomatik Corporation EKC33184
Encompass_Preferred
Human Cocaine-And Amphetamine-Regulated Transcript Protein (Cartpt) Elisa Kit for detecting Cocaine-And Amphetamine-Regulated Transcript Protein (Cartpt) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16568; CARTPT. Target Synonyms: CART; CART prepropeptide; CART (42-89); CART_HUMAN; Cartpt; Cocaine and amphetamine regulated transcript. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3875